N-(4-Chlorophenyl)-1,1,1-trifluoro-methanesulphonamide 95%

Lieferant: Apollo Scientific

PC520510-1G PC520510-5G
APOSPC520510-1GEA 280 EUR
APOSPC520510-1G APOSPC520510-5G
N-(4-Chlorophenyl)-1,1,1-trifluoro-methanesulphonamide 95%
N-(4-Chlorophenyl)-1,1,1-trifluoro-methanesulphonamide
Formel: C₇H₅ClF₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00856978
CAS-Nummer: 23384-04-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR