3',5'-Dimethyl-2,2,2-trifluoracetophenon

Lieferant: Apollo Scientific

Synonyme: 1-(3,5-Dimethylphenyl)-2,2,2-trifluoroethanone

PC520513-5G
APOSPC520513-5GEA 350 EUR
APOSPC520513-5G
3',5'-Dimethyl-2,2,2-trifluoracetophenon
3',5'-Dimethyl-2,2,2-trifluoracetophenon
Formel: C₁₀H₉F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD01319988
CAS-Nummer: 132719-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR