2-Methyl-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid 95%
Lieferant: Apollo Scientific
Synonyme:
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-5-carboxylic acid
PC520519-5G
APOSPC520519-5GEA
466
EUR
APOSPC520519-5G
2-Methyl-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid 95%
2-Methyl-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid
Formel:
C₆H₅F₃N₂O₂ Molecular Weight: 194,11 g/mol Schmelzpunkt: 130…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02090853 CAS-Nummer: 128694-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...