(6-Fluor-3-pyridinyl)methanol 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(6-Fluoropyridin-3-yl)methanol
PC520522-5G
APOSPC520522-5GEA
97.3
EUR
APOSPC520522-5G
(6-Fluor-3-pyridinyl)methanol 96%
(6-Fluor-3-pyridinyl)methanol
Formel:
C₆H₆FNO Molecular Weight: 127.1200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095186 CAS-Nummer: 39891-05-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...