2,4-Bis(trifluoromethyl)pyridine 95%
Lieferant: Apollo Scientific
PC520525-1G
APOSPC520525-1GEA
582
EUR
APOSPC520525-1G
2,4-Bis(trifluoromethyl)pyridine 95%
2,4-Bis(trifluoromethyl)pyridine
Formel:
C₇H₃F₆N Molecular Weight: 215,096 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093758 CAS-Nummer: 454-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...