4-(Trifluoromethyl)biphenyl 95%

Lieferant: Apollo Scientific

Synonyme: 4-(Trifluoromethyl)-1,1'-biphenyl

PC520526-25G
APOSPC520526-25GEA 524 EUR
APOSPC520526-25G
4-(Trifluoromethyl)biphenyl 95%
4-(Trifluoromethyl)biphenyl
Formel: C₁₃H₉F₃
Molecular Weight: 222,21 g/mol
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03412287
CAS-Nummer: 398-36-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR