3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid ≥95%

Lieferant: Apollo Scientific

Synonyme: RoflumilastRelated Compound D, 3-(Cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid

PC520539-25G
APOSPC520539-25GEA 208 EUR
APOSPC520539-25G
3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid ≥95%
3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid
Formel: C₁₂H₁₂F₂O₄
Molecular Weight: 258.221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04621687
CAS-Nummer: 162401-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR