2,5-Difluor-4-methoxybenzaldehyd 98%

Lieferant: Apollo Scientific

PC520540-5G
APOSPC520540-5GEA 127 EUR
APOSPC520540-5G
2,5-Difluor-4-methoxybenzaldehyd 98%
2,5-Difluoro-4-methoxybenzaldehyde
Formel: C₈H₆F₂O₂
Molecular Weight: 172.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06246882
CAS-Nummer: 879093-08-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR