3-(Difluoromethyl)-1-fluorobenzene 97%

Lieferant: Apollo Scientific

PC520546-25G PC520546-5G
APOSPC520546-25GEA 686 EUR
APOSPC520546-25G APOSPC520546-5G
3-(Difluoromethyl)-1-fluorobenzene 97%
3-(Difluoromethyl)-1-fluorobenzene
Formel: C₇H₅F₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06657983
CAS-Nummer: 26029-52-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR