2-(Trifluormethyl)benzylmercaptan 95%

Lieferant: Apollo Scientific

Synonyme: 2-(Trifluoromethyl)phenylmethanethiol

PC520547-5G
APOSPC520547-5GEA 222 EUR
APOSPC520547-5G
2-(Trifluormethyl)benzylmercaptan 95%
2-(Trifluormethyl)benzylmercaptan
Formel: C₈H₇F₃S
Lagertemperatur: Raumtemperatur
CAS-Nummer: 26039-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR