(6-Fluoro-2-pyridyl)methanol 97%

Lieferant: Apollo Scientific

PC520559-5G
APOSPC520559-5GEA 172 EUR
APOSPC520559-5G
(6-Fluoro-2-pyridyl)methanol 97%
(6-Fluoro-2-pyridyl)methanol
Formel: C₆H₆FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD07783827
CAS-Nummer: 315180-17-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR