[[4-(Trifluoromethyl)cyclohexyl]methyl]amine(cis- and trans- mixture) 97%

Lieferant: Apollo Scientific

PC520561-1G PC520561-500MG PC520561-5G
APOSPC520561-1GEA 292 EUR
APOSPC520561-1G APOSPC520561-500MG APOSPC520561-5G
[[4-(Trifluoromethyl)cyclohexyl]methyl]amine(cis- and trans- mixture) 97%
[[4-(Trifluoromethyl)cyclohexyl]methyl]amine(cis- and trans- mixture)
Formel: C₈H₁₄F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD07777219
CAS-Nummer: 361393-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR