2-Chlor-5-iodbenzotrifluorid 97%
Lieferant: Apollo Scientific
Synonyme:
1-Chloro-4-iodo-2-(trifluoromethyl)benzene
PC520562-100G
APOSPC520562-100GEA
121
EUR
APOSPC520562-100G
2-Chlor-5-iodbenzotrifluorid 97%
2-Chlor-5-iodbenzotrifluorid
Formel:
C₇H₃ClF₃I Molecular Weight: 306,4514 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09037801 CAS-Nummer: 260355-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...