1-Cyclohexyl-2,2,2-trifluoroethanamine 95%
Lieferant: Apollo Scientific
PC520565-5G
APOSPC520565-5GEA
1880
EUR
APOSPC520565-5G
1-Cyclohexyl-2,2,2-trifluoroethanamine 95%
1-Cyclohexyl-2,2,2-trifluoroethanamine
Formel:
C₈H₁₄F₃N Molecular Weight: 181,199 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07384540 CAS-Nummer: 1078726-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...