Potassium (2-cyanophenyl)trifluoroborate 97%

Lieferant: Apollo Scientific

PC520579-5G
APOSPC520579-5GEA 114 EUR
APOSPC520579-5G
Potassium (2-cyanophenyl)trifluoroborate 97%
Potassium (2-cyanophenyl)trifluoroborate
Formel: C₇H₄BF₃N·K
Lagertemperatur: Raumtemperatur
MDL: MFCD09952302
CAS-Nummer: 929038-12-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR