2,2'-Bis(trifluoromethyl)biphenyl 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC520582-10G
PC520582-5G
APOSPC520582-10GEA
1230
EUR
APOSPC520582-10G
APOSPC520582-5G
2,2'-Bis(trifluoromethyl)biphenyl 95%
2,2'-Bis(trifluoromethyl)biphenyl
Formel:
C₁₄H₈F₆ Molecular Weight: 290.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09836618 CAS-Nummer: 567-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...