2,2'-Bis(trifluoromethyl)biphenyl 95%

Lieferant: Apollo Scientific
Achtung

PC520582-10G PC520582-5G
APOSPC520582-10GEA 1230 EUR
APOSPC520582-10G APOSPC520582-5G
2,2'-Bis(trifluoromethyl)biphenyl 95%
2,2'-Bis(trifluoromethyl)biphenyl
Formel: C₁₄H₈F₆
Molecular Weight: 290.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09836618
CAS-Nummer: 567-15-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR