4-Brom-2-(trifluormethoxy)anisol 95%

Lieferant: Apollo Scientific

PC520587-10G PC520587-5G
APOSPC520587-10GEA 535 EUR
APOSPC520587-10G APOSPC520587-5G
4-Brom-2-(trifluormethoxy)anisol 95%
4-Brom-2-(trifluormethoxy)anisol
Formel: C₈H₆BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09907950
CAS-Nummer: 853771-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR