2-Bromo-1-(difluoromethoxy)-4-(trifluoromethyl)benzene 95%

Lieferant: Apollo Scientific

PC520590-10G
APOSPC520590-10GEA 2340 EUR
APOSPC520590-10G
2-Bromo-1-(difluoromethoxy)-4-(trifluoromethyl)benzene 95%
2-Bromo-1-(difluoromethoxy)-4-(trifluoromethyl)benzene
Formel: C₈H₄BrF₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD11036074
CAS-Nummer: 954236-03-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR