2-Chloro-6-methyl-3-(trifluoromethyl)pyridine 97%

Lieferant: Apollo Scientific

PC520613-10G PC520613-1G PC520613-5G
APOSPC520613-10GEA 1160 EUR
APOSPC520613-10G APOSPC520613-1G APOSPC520613-5G
2-Chloro-6-methyl-3-(trifluoromethyl)pyridine 97%
2-Chloro-6-methyl-3-(trifluoromethyl)pyridine
Formel: C₇H₅ClF₃N
Molecular Weight: 195.57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226678
CAS-Nummer: 1099597-74-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR