4-Bromo-1-fluoro-2-(2,2,2-trifluoroethyl)benzene 95%
Lieferant: Apollo Scientific
PC520617-5G
APOSPC520617-5GEA
919
EUR
APOSPC520617-5G
4-Bromo-1-fluoro-2-(2,2,2-trifluoroethyl)benzene 95%
4-Bromo-1-fluoro-2-(2,2,2-trifluoroethyl)benzene
Formel:
C₈H₅BrF₄ Molecular Weight: 257,023 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226694 CAS-Nummer: 1186195-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...