2-Methyl-4-nitrobenzotrifluoride 97%

Lieferant: Apollo Scientific

Synonyme: 2-Methyl-4-nitro-1-(trifluoromethyl)benzene

PC520626-5G
APOSPC520626-5GEA 408 EUR
APOSPC520626-5G
2-Methyl-4-nitrobenzotrifluoride 97%
2-Methyl-4-nitrobenzotrifluoride
Formel: C₈H₆F₃NO₂
Molecular Weight: 205,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226539
CAS-Nummer: 1960-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR