3-(N-Boc-amino)benzotrifluorid ≥95%

Lieferant: Apollo Scientific

Synonyme: (3-Trifluoromethylphenyl)-carbamic acid tert-butyl ester

PC520634-1G
APOSPC520634-1GEA 268 EUR
APOSPC520634-1G
3-(N-Boc-amino)benzotrifluorid ≥95%
3-(N-Boc-amino)benzotrifluorid
Formel: C₁₂H₁₄F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11040633
CAS-Nummer: 109134-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR