1-(1,1-Difluoroethyl)-2,3-difluorobenzene 95%

Lieferant: Apollo Scientific

PC520640-5G
APOSPC520640-5GEA 1630 EUR
APOSPC520640-5G
1-(1,1-Difluoroethyl)-2,3-difluorobenzene 95%
1-(1,1-Difluoroethyl)-2,3-difluorobenzene
Formel: C₈H₆F₄
Lagertemperatur: Raumtemperatur
MDL: MFCD11845810
CAS-Nummer: 1138445-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR