1-Chloro-4-(1,1-difluoroethyl)-2,5-difluorobenzene 95%

Lieferant: Apollo Scientific

PC520648-5G
APOSPC520648-5GEA 605 EUR
APOSPC520648-5G
1-Chloro-4-(1,1-difluoroethyl)-2,5-difluorobenzene 95%
1-Chloro-4-(1,1-difluoroethyl)-2,5-difluorobenzene
Formel: C₈H₅ClF₄
Molecular Weight: 212.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226702
CAS-Nummer: 1138444-89-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR