1-Fluoro-4-isopropenyl-2-(trifluoromethyl)benzene 95%

Lieferant: Apollo Scientific

PC520652-5G
APOSPC520652-5GEA 408 EUR
APOSPC520652-5G
1-Fluoro-4-isopropenyl-2-(trifluoromethyl)benzene 95%
1-Fluoro-4-isopropenyl-2-(trifluoromethyl)benzene
Formel: C₁₀H₈F₄
Lagertemperatur: Raumtemperatur
MDL: MFCD11226534
CAS-Nummer: 367-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR