3'-Brom-5'-(trifluormethyl)acetophenon 99%

Lieferant: Apollo Scientific

Synonyme: Ethanone, 1-[3-bromo-5-(trifluoromethyl)phenyl]-, 1-[3-Bromo-5-(trifluoromethyl)phenyl]ethanone

PC520654-1G PC520654-5G
APOSPC520654-1GEA 70.1 EUR
APOSPC520654-1G APOSPC520654-5G
3'-Brom-5'-(trifluormethyl)acetophenon 99%
3'-Brom-5'-(trifluormethyl)acetophenon
Formel: C₉H₆BrF₃O
Molecular Weight: 267.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847339
CAS-Nummer: 154259-25-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR