(3-Fluoropyridin-4-yl)methanol 99%

Lieferant: Apollo Scientific
Achtung

PC520662-1G PC520662-5G
APOSPC520662-1GEA 0 EUR
APOSPC520662-1G APOSPC520662-5G
(3-Fluoropyridin-4-yl)methanol 99%
(3-Fluoropyridin-4-yl)methanol
Formel: C₆H₆FNO
Lagertemperatur: Kühlschrank
MDL: MFCD11656367
CAS-Nummer: 870063-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR