1,2-Dichloro-4,5-bis-(trifluoromethyl)benzene 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC520674-10G
PC520674-1G
PC520674-5G
APOSPC520674-10GEA
1230
EUR
APOSPC520674-10G
APOSPC520674-1G
APOSPC520674-5G
1,2-Dichloro-4,5-bis-(trifluoromethyl)benzene 95%
1,2-Dichloro-4,5-bis(trifluoromethyl)benzene
Formel:
C₈H₂Cl₂F₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100166 CAS-Nummer: 320-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...