1,2-Dichloro-4,5-bis(trifluoromethyl)benzene 95%

Lieferant: Apollo Scientific
Achtung

PC520674-10G PC520674-1G PC520674-5G
APOSPC520674-10GEA 0 EUR
APOSPC520674-10G APOSPC520674-1G APOSPC520674-5G
1,2-Dichloro-4,5-bis(trifluoromethyl)benzene 95%
1,2-Dichloro-4,5-bis(trifluoromethyl)benzene
Formel: C₈H₂Cl₂F₆
Lagertemperatur: Raumtemperatur
MDL: MFCD11100166
CAS-Nummer: 320-22-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR