1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulphonate 90%
Lieferant: Apollo Scientific
Synonyme:
1-[2-(2,4-Difluorophenyl)-oxiranyl]-methyl-1H-1,2,4-triazolemonomethanesulphonate, 2-(2,4-Difluorophenyl)-2,3-epoxy-1-(1H-1,2,4-triazol-1-yl)propane methanesulphonate salt, 2-[[(2,4-Difluorophenyl)-2-oxiranyl]methyl]-1H-1,2,4-triazole methanesulphonate
PC520675-25G
APOSPC520675-25GEA
224
EUR
APOSPC520675-25G
1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulphonate 90%
1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulphonate
Formel:
C₁₂H₁₃F₂N₃O₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD11111978 CAS-Nummer: 86386-77-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...