3-(Trifluoromethoxy)phenylmethanesulphonyl chloride 97%
Lieferant: Apollo Scientific
PC520678-1G
PC520678-5G
APOSPC520678-1GEA
803
EUR
APOSPC520678-1G
APOSPC520678-5G
3-(Trifluoromethoxy)phenylmethanesulphonyl chloride 97%
3-(Trifluoromethoxy)phenylmethanesulphonyl chloride
Formel:
C₈H₆ClF₃O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD11976109 CAS-Nummer: 1263274-68-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...