1,3-Difluoro-2,4-dimethoxy-5-nitrobenzene 95%
Lieferant: Apollo Scientific
PC520686-1G
PC520686-5G
APOSPC520686-1GEA
466
EUR
APOSPC520686-1G
APOSPC520686-5G
1,3-Difluoro-2,4-dimethoxy-5-nitrobenzene 95%
1,3-Difluoro-2,4-dimethoxy-5-nitrobenzene
Formel:
C₈H₇F₂NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12547866 CAS-Nummer: 195136-63-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...