1,3-Difluoro-2,4-dimethoxy-5-nitrobenzene 95%

Lieferant: Apollo Scientific

PC520686-1G PC520686-5G
APOSPC520686-1GEA 466 EUR
APOSPC520686-1G APOSPC520686-5G
1,3-Difluoro-2,4-dimethoxy-5-nitrobenzene 95%
1,3-Difluoro-2,4-dimethoxy-5-nitrobenzene
Formel: C₈H₇F₂NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD12547866
CAS-Nummer: 195136-63-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR