5-Bromo-3-fluoro-2-(hydroxymethyl)pyridine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Bromo-3-fluoropyridin-2-yl)methanol

PC52069-250MG
APOSPC52069-250MGEA 92.8 EUR
APOSPC52069-250MG
5-Bromo-3-fluoro-2-(hydroxymethyl)pyridine 97%
5-Bromo-3-fluoro-2-(hydroxymethyl)pyridine
Formel: C₆H₅BrFNO
Molecular Weight: 206,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16607004
CAS-Nummer: 1206968-92-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR