2-Chlor-4-fluor-3-methylbenzonitril 95%

Lieferant: Apollo Scientific

PC520690-5G
APOSPC520690-5GEA 84.1 EUR
APOSPC520690-5G
2-Chlor-4-fluor-3-methylbenzonitril 95%
2-Chlor-4-fluor-3-methylbenzonitril
Formel: C₈H₅ClFN
Molecular Weight: 169,5834 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13193544
CAS-Nummer: 796600-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR