2-(Trifluoromethyl)cyclopropanamine hydrochloride 95%

Lieferant: Apollo Scientific

PC520694-1G PC520694-250MG PC520694-500MG
APOSPC520694-1GEA 1150 EUR
APOSPC520694-1G APOSPC520694-250MG APOSPC520694-500MG
2-(Trifluoromethyl)cyclopropanamine hydrochloride 95%
2-(Trifluoromethyl)cyclopropanamine hydrochloride
Formel: C₄H₆F₃N·ClH
Molecular Weight: 161.553 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13806389
CAS-Nummer: 1172072-62-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR