4-Fluoro-2-methoxybenzenesulphonyl chloride 95%

Lieferant: Apollo Scientific

PC52070-1G PC52070-250MG
APOSPC52070-1GEA 899 EUR
APOSPC52070-1G APOSPC52070-250MG
4-Fluoro-2-methoxybenzenesulphonyl chloride 95%
4-Fluoro-2-methoxybenzenesulphonyl chloride
Formel: C₇H₆ClFO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD13185596
CAS-Nummer: 1214377-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR