4-(Trifluoromethyl)cyclohexanecarboxylic acid methyl ester 97%
Lieferant: Apollo Scientific
Synonyme:
Methyl 4-(trifluoromethyl)cyclohexane-1-carboxylate
PC520700-25G
PC520700-5G
APOSPC520700-25GEA
815
EUR
APOSPC520700-25G
APOSPC520700-5G
4-(Trifluoromethyl)cyclohexanecarboxylic acid methyl ester 97%
4-(Trifluoromethyl)cyclohexanecarboxylic acid methyl ester
Formel:
C₉H₁₃F₃O₂ Molecular Weight: 210.2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14525503 CAS-Nummer: 1204296-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...