4-(Trifluoromethyl)cyclohexanecarboxylic acid methyl ester 97%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-(trifluoromethyl)cyclohexane-1-carboxylate

PC520700-25G PC520700-5G
APOSPC520700-25GEA 815 EUR
APOSPC520700-25G APOSPC520700-5G
4-(Trifluoromethyl)cyclohexanecarboxylic acid methyl ester 97%
4-(Trifluoromethyl)cyclohexanecarboxylic acid methyl ester
Formel: C₉H₁₃F₃O₂
Molecular Weight: 210.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14525503
CAS-Nummer: 1204296-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR