1-Chloro-2-(difluoromethyl)-4-(trifluoromethyl)benzene 95%

Lieferant: Apollo Scientific

PC520708-5G
APOSPC520708-5GEA 686 EUR
APOSPC520708-5G
1-Chloro-2-(difluoromethyl)-4-(trifluoromethyl)benzene 95%
1-Chloro-2-(difluoromethyl)-4-(trifluoromethyl)benzene
Formel: C₈H₄ClF₅
Lagertemperatur: Raumtemperatur
MDL: MFCD14698551
CAS-Nummer: 1214383-34-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR