2-Difluoromethylbenzoic acid methyl ester 95%

Lieferant: Apollo Scientific

PC520720-10G PC520720-1G PC520720-5G
APOSPC520720-10GEA 975 EUR
APOSPC520720-10G APOSPC520720-1G APOSPC520720-5G
2-Difluoromethylbenzoic acid methyl ester 95%
2-Difluoromethylbenzoic acid methyl ester
Formel: C₉H₈F₂O₂
Molecular Weight: 186.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16140192
CAS-Nummer: 1018678-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR