1-Bromo-4-(1,1-difluoropropyl)benzene 95%

Lieferant: Apollo Scientific

PC520724-1G
APOSPC520724-1GEA 579 EUR
APOSPC520724-1G
1-Bromo-4-(1,1-difluoropropyl)benzene 95%
1-Bromo-4-(1,1-difluoropropyl)benzene
Formel: C₉H₉BrF₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16140148
CAS-Nummer: 162783-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR