2,2-Difluoro-1-phenylcyclopropanecarboxylic acid 97%

Lieferant: Apollo Scientific

PC520742-1G PC520742-500MG PC520742-5G
APOSPC520742-1GEA 361 EUR
APOSPC520742-1G APOSPC520742-500MG APOSPC520742-5G
2,2-Difluoro-1-phenylcyclopropanecarboxylic acid 97%
2,2-Difluoro-1-phenylcyclopropanecarboxylic acid
Formel: C₁₀H₈F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18434522
CAS-Nummer: 156021-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR