1-(2-Fluorobenzyl)-1H-pyrazol-4-ylamine 95%

Lieferant: Apollo Scientific

Synonyme: 1-(2-Fluorobenzyl)-1H-pyrazol-4-amine hydrochloride

PC520744-1G
APOSPC520744-1GEA 442 EUR
APOSPC520744-1G
1-(2-Fluorobenzyl)-1H-pyrazol-4-ylamine 95%
1-(2-Fluorobenzyl)-1H-pyrazol-4-ylamine
Formel: C₁₀H₁₀FN₃
Molecular Weight: 191,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02253989
CAS-Nummer: 925634-52-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR