2-Chloro-6-iodo-4-(trifluoromethyl)aniline 97%

Lieferant: Apollo Scientific

PC520752-10G PC520752-1G PC520752-5G
APOSPC520752-10GEA 1040 EUR
APOSPC520752-10G APOSPC520752-1G APOSPC520752-5G
2-Chloro-6-iodo-4-(trifluoromethyl)aniline 97%
2-Chloro-6-iodo-4-(trifluoromethyl)aniline
Formel: C₇H₄ClF₃IN
Lagertemperatur: Raumtemperatur
MDL: MFCD20492612
CAS-Nummer: 1065102-88-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR