3-(Difluoromethyl)-2,6-difluoro-pyridine 97%

Lieferant: Apollo Scientific
Achtung

PC520761-10G PC520761-1G PC520761-5G
APOSPC520761-10GEA 415 EUR
APOSPC520761-10G APOSPC520761-1G APOSPC520761-5G
3-(Difluoromethyl)-2,6-difluoro-pyridine 97%
3-(Difluoromethyl)-2,6-difluoro-pyridine
Formel: C₆H₃F₄N
Molecular Weight: 165.09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22199312
CAS-Nummer: 85396-63-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR