2,6-Dichloro-3-(difluoromethyl)-5-fluoropyridine 97%

Lieferant: Apollo Scientific

PC520763-5G
APOSPC520763-5GEA 1350 EUR
APOSPC520763-5G
2,6-Dichloro-3-(difluoromethyl)-5-fluoropyridine 97%
2,6-Dichloro-3-(difluoromethyl)-5-fluoropyridine
Formel: C₆H₂Cl₂F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD22199308
CAS-Nummer: 1374659-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR