1-(Difluoromethoxy)-2,3-difluorobenzene 95%
Lieferant: Apollo Scientific
PC520766-10G
APOSPC520766-10GEA
2310
EUR
APOSPC520766-10G
1-(Difluoromethoxy)-2,3-difluorobenzene 95%
1-(Difluoromethoxy)-2,3-difluorobenzene
Formel:
C₇H₄F₄O Molecular Weight: 180,1 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22580881 CAS-Nummer: 1404194-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...