1-(Difluoromethoxy)-2-fluoro-3-(trifluoromethyl)benzene 95%
Lieferant: Apollo Scientific
PC520768-10G
APOSPC520768-10GEA
2310
EUR
APOSPC520768-10G
1-(Difluoromethoxy)-2-fluoro-3-(trifluoromethyl)benzene 95%
1-(Difluoromethoxy)-2-fluoro-3-(trifluoromethyl)benzene
Formel:
C₈H₄F₆O Lagertemperatur: Raumtemperatur |
MDL:
MFCD22580904 CAS-Nummer: 1417569-57-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...